Assay Design
Gene Transcripts
- Gene Symbol
- Alpk3
- Entrez Gene ID
-
116904
- Gene Name
- alpha-kinase 3
- Gene Aliases
-
AW319487, D330016D04, MAK, Midori, mKIAA1330
- Chromosome Location
-
Chr.7: 81057600 - 81105612 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF338872.1 |
- |
1 - 2 |
172 |
108 |
View Full Product Details