Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109758.1 | NP_001103228.1 | 6 - 7 | 1332 | 68 |
NM_007529.2 | NP_031555.2 | 6 - 7 | 1332 | 68 | |
GenBank mRNA | AK144405.1 | - | 6 - 7 | 1132 | 68 |
BC052032.1 | - | 6 - 7 | 1127 | 68 | |
X87096.1 | - | 6 - 7 | 1208 | 68 |