Assay Design



Gene Transcripts

Gene Symbol
Pcolce
Entrez Gene ID
18542
Gene Name
procollagen C-endopeptidase enhancer protein
Gene Aliases
Astt-2, Astt2, P14, PCPE-1
Chromosome Location
Chr.5: 137605105 - 137611899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008788.2 NP_032814.2 1 - 2 174 102
GenBank mRNA AB008548.1 - 1 - 2 98 102
AK142615.1 - 1 - 2 103 102
AK144392.1 - 1 - 2 175 102
AK146829.1 - 1 - 2 175 102
AK159430.1 - 1 - 2 162 102
AK159940.1 - 1 - 2 167 102
AK162524.1 - 1 - 2 178 102
X57337.1 - 1 - 2 141 102

View Full Product Details