Assay Design



Gene Transcripts

Gene Symbol
Pde6b
Entrez Gene ID
18587
Gene Name
phosphodiesterase 6B, cGMP, rod receptor, beta polypeptide
Gene Aliases
Pdeb, r, rd, rd-1, rd1, rd10
Chromosome Location
Chr.5: 108388373 - 108431743 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008806.2 NP_032832.2 14 - 15 1903 69
GenBank mRNA AK039397.1 - 14 - 15 1615 69
AK044364.1 - 14 - 15 1846 69
BC048247.1 - 14 - 15 1882 69
BC129923.1 - 14 - 15 1833 69
BC129924.1 - 14 - 15 1832 69
X55968.1 - 14 - 15 1836 69
X60133.1 - 14 - 15 69
X87952.1 - 14 - 15 69

View Full Product Details