Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008894.2 | NP_032920.2 | 5 - 6 | 697 | 72 |
GenBank mRNA | AK013901.1 | - | 5 - 6 | 72 | |
BC003342.1 | - | 5 - 6 | 663 | 72 | |
CB205181.1 | - | 2 - 3 | 233 | 72 | |
Z72486.1 | - | 5 - 6 | 658 | 72 |