Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008247.3 | NP_032273.1 | 4 - 5 | 981 | 74 |
NM_008903.2 | NP_032929.1 | 4 - 5 | 978 | 74 | |
GenBank mRNA | AK077275.1 | - | 4 - 5 | 865 | 74 |
AK205792.1 | - | 2 - 3 | 164 | 74 | |
AY247795.1 | - | 4 - 5 | 603 | 74 | |
AY247796.1 | - | 4 - 5 | 606 | 74 | |
BC061161.1 | - | 4 - 5 | 966 | 74 | |
CD742481.1 | - | 2 - 3 | 74 | ||
D84376.1 | - | 4 - 5 | 832 | 74 |