Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010726.2 | NP_034856.1 | 5 - 6 | 572 | 81 |
GenBank mRNA | AF023463.1 | - | 5 - 6 | 537 | 81 |
AK146753.1 | - | 5 - 6 | 557 | 81 | |
AK151372.1 | - | 5 - 6 | 520 | 81 | |
AK164495.1 | - | 5 - 6 | 523 | 81 | |
AK165412.1 | - | 5 - 6 | 516 | 81 | |
AK168648.1 | - | 5 - 6 | 542 | 81 | |
AK169213.1 | - | 5 - 6 | 514 | 81 | |
BC002018.1 | - | 5 - 6 | 549 | 81 | |
D88670.1 | - | 5 - 6 | 532 | 81 |