Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010947.3 | NP_035077.1 | 3 - 4 | 1666 | 82 |
GenBank mRNA | AF128866.1 | - | 3 - 4 | 1506 | 82 |
AF149094.1 | - | 3 - 4 | 1302 | 82 | |
AF152418.1 | - | 3 - 4 | 1421 | 82 | |
AK031199.1 | - | 3 - 4 | 1811 | 82 | |
AK141856.1 | - | 3 - 4 | 1577 | 82 | |
BC053038.1 | - | 3 - 4 | 1758 | 82 | |
BC094362.1 | - | 3 - 4 | 1666 | 82 |