Assay Design



Gene Transcripts

Gene Symbol
Nudc
Entrez Gene ID
18221
Gene Name
nuclear distribution gene C homolog (Aspergillus)
Gene Aliases
SIG-92, Silg92
Chromosome Location
Chr.4: 133532542 - 133546027 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010948.3 NP_035078.1 2 - 3 301 89
GenBank mRNA AK012321.1 - 2 - 3 271 89
AK012777.1 - 2 - 3 250 89
AK032673.1 - 2 - 3 258 89
AK146323.1 - 2 - 3 271 89
BC011253.1 - 2 - 3 236 89
BY065900.1 - 2 - 3 272 90
DV652222.1 - 2 - 3 301 89
X81443.1 - 2 - 3 238 89
Y15522.1 - 2 - 3 162 89

View Full Product Details