Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011198.3 | NP_035328.2 | 8 - 9 | 1358 | 90 |
GenBank mRNA | AK049923.1 | - | 8 - 9 | 1349 | 90 |
AK144726.1 | - | 8 - 9 | 1383 | 90 | |
AK144956.1 | - | 8 - 9 | 1381 | 90 | |
AK149820.1 | - | 8 - 9 | 1383 | 90 | |
AK166221.1 | - | 8 - 9 | 1381 | 90 | |
AK172161.1 | - | 8 - 9 | 1383 | 90 | |
BC052900.1 | - | 8 - 9 | 1225 | 90 | |
M64291.1 | - | 8 - 9 | 1382 | 90 | |
M88242.1 | - | 8 - 9 | 1358 | 90 | |
M94967.1 | - | 8 - 9 | 1383 | 90 |