Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017463.2 | NP_059491.1 | 1 - 2 | 306 | 116 |
GenBank mRNA | AF020198.1 | - | 1 - 2 | 308 | 116 |
AK134357.1 | - | 1 - 2 | 227 | 116 | |
AK159623.1 | - | 1 - 2 | 515 | 116 | |
BC087615.1 | - | 1 - 2 | 286 | 116 |