Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163379.1 | NP_001156851.1 | 5 - 6 | 920 | 68 |
NM_018862.3 | NP_061350.3 | 5 - 6 | 672 | 68 | |
GenBank mRNA | AB005623.1 | - | 5 - 6 | 918 | 68 |
AK086281.1 | - | 5 - 6 | 862 | 68 | |
BC009651.1 | - | 5 - 6 | 672 | 68 |