Assay Design



Gene Transcripts

Gene Symbol
Paxip1
Entrez Gene ID
55982
Gene Name
PAX interacting (with transcription-activation domain) protein 1
Gene Aliases
D5Ertd149e, PTIP
Chromosome Location
Chr.5: 27740080 - 27792079 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018878.3 NP_061366.2 6 - 7 1325 93
GenBank mRNA AF104261.1 - 6 - 7 1325 93
AK144606.1 - 6 - 7 1187 93
AK148141.1 - 6 - 7 1196 93
BC066014.1 - 6 - 7 1114 93

View Full Product Details