Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018878.3 | NP_061366.2 | 7 - 8 | 2031 | 78 |
GenBank mRNA | AF104261.1 | - | 7 - 8 | 2031 | 78 |
AK144606.1 | - | 7 - 8 | 1893 | 78 | |
AK148141.1 | - | 7 - 8 | 1901 | 78 | |
BC066014.1 | - | 7 - 8 | 1820 | 78 |