Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019458.3 | NP_062331.2 | 12 - 13 | 1307 | 64 |
GenBank mRNA | AB041615.1 | - | 12 - 13 | 1325 | 64 |
AK017762.1 | - | 12 - 13 | 1377 | 64 | |
AK134857.1 | - | 12 - 13 | 1320 | 64 | |
AK148538.1 | - | 12 - 13 | 1342 | 64 | |
AK151665.1 | - | 12 - 13 | 1353 | 64 | |
AK152778.1 | - | 12 - 13 | 1323 | 64 | |
AK152836.1 | - | 12 - 13 | 1353 | 64 | |
BC010317.1 | - | 11 - 12 | 1351 | 64 | |
BC029843.1 | - | 12 - 13 | 1307 | 64 | |
BC083337.1 | - | 12 - 13 | 1288 | 64 |