Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085472.2 | NP_001078941.2 | 7 - 8 | 692 | 91 |
NM_001085473.2 | NP_001078942.2 | 13 - 14 | 3017 | 91 | |
NM_001242605.1 | NP_001229534.1 | 6 - 7 | 756 | 91 | |
NM_001242606.1 | NP_001229535.1 | 6 - 7 | 717 | 91 | |
NM_019567.3 | NP_062513.3 | 7 - 8 | 821 | 91 | |
NM_023190.3 | NP_075679.2 | 13 - 14 | 3053 | 91 | |
GenBank mRNA | AF124725.1 | - | 7 - 8 | 752 | 91 |
AF124729.1 | - | 6 - 7 | 687 | 91 | |
AF168782.1 | - | 13 - 14 | 3054 | 91 | |
AK012099.1 | - | 7 - 8 | 685 | 91 | |
AK030449.1 | - | 13 - 14 | 2747 | 91 | |
AK122342.1 | - | 13 - 14 | 2688 | 91 | |
BC019127.1 | - | 6 - 7 | 741 | 91 | |
BC052755.1 | - | 6 - 7 | 677 | 91 | |
BC075691.1 | - | 13 - 14 | 2702 | 91 | |
BC094217.1 | - | 6 - 7 | 699 | 91 | |
BY711288.1 | - | 7 - 8 | 685 | 91 |