Assay Design



Gene Transcripts

Gene Symbol
Hapln2
Entrez Gene ID
73940
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
4930401E20Rik, Bral1
Chromosome Location
Chr.3: 88020558 - 88027511 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022031.2 NP_071314.1 5 - 6 792 84
GenBank mRNA AB049055.1 - 5 - 6 721 84
BC023911.1 - 4 - 5 1073 84
BC138584.1 - 4 - 5 677 84
BC138585.1 - 4 - 5 677 84

View Full Product Details