Assay Design



Gene Transcripts

Gene Symbol
Ndst4
Entrez Gene ID
64580
Gene Name
N-deacetylase/N-sulfotransferase (heparin glucosaminyl) 4
Gene Aliases
4930439H17Rik
Chromosome Location
Chr.3: 125404076 - 125728351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022565.2 NP_072087.2 9 - 10 2592 121
GenBank mRNA AB036838.1 - 8 - 9 2042 121
AK132938.1 - 4 - 5 747 121

View Full Product Details