Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025364.2 | NP_079640.1 | 4 - 5 | 417 | 114 |
GenBank mRNA | AK003453.1 | - | 4 - 5 | 269 | 114 |
AK018773.1 | - | 4 - 5 | 280 | 114 | |
AK088266.1 | - | 4 - 5 | 266 | 114 | |
AK146248.1 | - | 4 - 5 | 295 | 114 | |
AK212934.1 | - | 5 - 6 | 301 | 114 | |
BC027510.1 | - | 4 - 5 | 256 | 114 | |
BC094393.1 | - | 4 - 5 | 253 | 114 | |
BC157909.1 | - | 4 - 5 | 281 | 114 | |
BC158023.1 | - | 4 - 5 | 281 | 114 | |
BC172063.1 | - | 4 - 5 | 278 | 114 |