Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025376.3 | NP_079652.1 | 1 - 2 | 276 | 68 |
GenBank mRNA | AK003298.1 | - | 1 - 2 | 278 | 68 |
AK004477.1 | - | 1 - 2 | 245 | 68 | |
BB839715.1 | - | 1 - 2 | 278 | 68 | |
BC062145.1 | - | 1 - 2 | 202 | 68 |