Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025443.2 | NP_079719.1 | 2 - 3 | 428 | 80 |
GenBank mRNA | AF349950.1 | - | 2 - 3 | 411 | 80 |
AK007324.1 | - | 2 - 3 | 393 | 80 | |
AK009146.1 | - | 2 - 3 | 395 | 80 | |
AK146152.1 | - | 2 - 3 | 422 | 80 | |
AK165942.1 | - | 2 - 3 | 389 | 80 | |
AK168177.1 | - | 2 - 3 | 422 | 80 | |
BC020037.1 | - | 2 - 3 | 411 | 80 |