Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110809.2 | NP_001104279.1 | 7 - 8 | 600 | 97 |
NM_001286857.1 | NP_001273786.1 | 7 - 8 | 1214 | 97 | |
NM_025663.4 | NP_079939.3 | 8 - 9 | 735 | 97 | |
GenBank mRNA | AK011617.1 | - | 7 - 8 | 599 | 97 |
AK017310.1 | - | 7 - 8 | 550 | 97 | |
AK018306.1 | - | 7 - 8 | 574 | 97 | |
AK169111.1 | - | 7 - 8 | 589 | 97 | |
BC002265.1 | - | 7 - 8 | 571 | 97 | |
BC019490.1 | - | 8 - 9 | 717 | 97 | |
BC080676.1 | - | 7 - 8 | 1192 | 97 |