Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025799.4 | NP_080075.2 | 2 - 3 | 539 | 93 |
GenBank mRNA | AK002675.1 | - | 2 - 3 | 481 | 93 |
AK008653.1 | - | 2 - 3 | 484 | 93 | |
AK017443.1 | - | 2 - 3 | 510 | 93 | |
AK030320.1 | - | 2 - 3 | 541 | 93 | |
AK035607.1 | - | 2 - 3 | 420 | 93 | |
AK044790.1 | - | 2 - 3 | 396 | 93 | |
AK146295.1 | - | 2 - 3 | 468 | 93 | |
AK151204.1 | - | 2 - 3 | 400 | 93 | |
AK159288.1 | - | 2 - 3 | 485 | 93 | |
BC004039.1 | - | 2 - 3 | 490 | 93 | |
BC018181.1 | - | 2 - 3 | 499 | 93 | |
BC094449.1 | - | 2 - 3 | 454 | 93 |