Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025822.3 | NP_080098.1 | 1 - 2 | 130 | 113 |
GenBank mRNA | AK004742.2 | - | 1 - 2 | 84 | 113 |
AK012100.1 | - | 1 - 2 | 131 | 113 | |
AK043731.1 | - | 1 - 2 | 119 | 113 | |
AK045433.1 | - | 1 - 2 | 117 | 113 | |
AK168539.1 | - | 1 - 2 | 101 | 113 | |
AK169340.1 | - | 1 - 2 | 84 | 113 | |
BC083123.1 | - | 1 - 2 | 81 | 113 |