Assay Design



Gene Transcripts

Gene Symbol
Serpinb11
Entrez Gene ID
66957
Gene Name
serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 11
Gene Aliases
2310046M08Rik, AU015399
Chromosome Location
Chr.1: 107362314 - 107380475 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025867.2 NP_080143.1 1 - 2 42 117
GenBank mRNA AK009003.1 - 1 - 2 44 117
AK009855.1 - 1 - 2 43 117

View Full Product Details