Assay Design
Gene Transcripts
- Gene Symbol
- Pbld2
- Entrez Gene ID
-
67307
- Gene Name
- phenazine biosynthesis-like protein domain containing 2
- Gene Aliases
-
3110049J23Rik, Mawbp
- Chromosome Location
-
Chr.10: 63024049 - 63058813 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026085.2 |
NP_080361.1 |
1 - 2 |
222 |
59 |
GenBank mRNA |
AK014198.1 |
- |
1 - 2 |
223 |
59 |
View Full Product Details