Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007619.2 | NP_031645.2 | 10 - 11 | 1714 | 81 |
GenBank mRNA | AK029826.1 | - | 10 - 11 | 1717 | 81 |
AK153915.1 | - | 10 - 11 | 1666 | 81 | |
BC125285.1 | - | 10 - 11 | 1629 | 81 | |
X57111.1 | - | 10 - 11 | 1601 | 81 |