Assay Design



Gene Transcripts

Gene Symbol
Ncan
Entrez Gene ID
13004
Gene Name
neurocan
Gene Aliases
C230035B04, Cspg3, Cspg3-rs, Tgfbit
Chromosome Location
Chr.8: 70093082 - 70120870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007789.3 NP_031815.2 2 - 3 200 75
GenBank mRNA BC065118.1 - 2 - 3 166 75
CX202471.1 - 2 - 3 200 75
X84727.1 - 2 - 3 162 75

View Full Product Details