Assay Design



Gene Transcripts

Gene Symbol
Galc
Entrez Gene ID
14420
Gene Name
galactosylceramidase
Gene Aliases
2310068B06Rik, A930008M05Rik, AW212969, AW413532, Gacy, twi, twitcher
Chromosome Location
Chr.12: 98202300 - 98259461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK040189.1 - 3 - 4 949 96
AK077475.1 - 3 - 4 304 96
AK154760.1 - 3 - 4 465 96
BC065140.1 - 3 - 4 338 96
BC086671.1 - 3 - 4 342 96
D38557.1 - 3 - 4 412 96

View Full Product Details