Assay Design



Gene Transcripts

Gene Symbol
Gbas
Entrez Gene ID
14467
Gene Name
glioblastoma amplified sequence
Gene Aliases
AV006093, Nipsnap2
Chromosome Location
Chr.5: 129725044 - 129758327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008095.4 NP_032121.3 6 - 7 643 72
GenBank mRNA AJ001261.1 - 6 - 7 621 72
AK170336.1 - 6 - 7 589 72
BC055893.1 - 6 - 7 592 72

View Full Product Details