Assay Design



Gene Transcripts

Gene Symbol
Anapc1
Entrez Gene ID
17222
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24
Chromosome Location
Chr.2: 128610083 - 128687410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008569.2 NP_032595.2 44 - 45 5555 91
GenBank mRNA AK034858.1 - 12 - 13 1086 91
AK090134.1 - 44 - 45 5537 91
BC139002.1 - 44 - 45 5536 91
BC145492.1 - 44 - 45 5536 91
X80169.1 - 44 - 45 5540 91

View Full Product Details