Assay Design



Gene Transcripts

Gene Symbol
Mmp15
Entrez Gene ID
17388
Gene Name
matrix metallopeptidase 15
Gene Aliases
AI503551, MT2-MMP
Chromosome Location
Chr.8: 95352337 - 95374293 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008609.3 NP_032635.1 1 - 2 678 129
GenBank mRNA BC047278.1 - 1 - 2 678 129
BC057952.1 - 1 - 2 248 129
D86332.1 - 1 - 2 174 129

View Full Product Details