Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008609.3 | NP_032635.1 | 7 - 8 | 1826 | 69 |
GenBank mRNA | AY622975.1 | - | 2 - 3 | 233 | 69 |
BC047278.1 | - | 7 - 8 | 1826 | 69 | |
BC057952.1 | - | 7 - 8 | 1396 | 69 | |
D86332.1 | - | 7 - 8 | 1322 | 69 |