Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008624.3 | NP_032650.1 | 2 - 3 | 617 | 69 |
GenBank mRNA | AB004879.1 | - | 2 - 3 | 601 | 69 |
AF031159.1 | - | 1 - 2 | 197 | 69 | |
AF043581.1 | - | 1 - 2 | 215 | 69 | |
AK050469.1 | - | 2 - 3 | 461 | 69 | |
AK164066.1 | - | 2 - 3 | 261 | 69 | |
BC024389.1 | - | 2 - 3 | 334 | 69 |