Assay Design



Gene Transcripts

Gene Symbol
Meis3
Entrez Gene ID
17537
Gene Name
Meis homeobox 3
Gene Aliases
AI573393, Mrg2
Chromosome Location
Chr.7: 16174849 - 16186510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC003762.1 - 12 - 13 1262 68
BC118513.1 - 12 - 13 1223 68
U57344.1 - 12 - 13 1289 68

View Full Product Details