Assay Design
Gene Transcripts
- Gene Symbol
- Map2
- Entrez Gene ID
-
17756
- Gene Name
- microtubule-associated protein 2
- Gene Aliases
-
G1-397-34, MAP-2, Mtap-2, Mtap2, repro4
- Chromosome Location
-
Chr.1: 66175246 - 66442583 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M21041.1 |
- |
6 - 7 |
666 |
65 |
View Full Product Details