Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009105.4 | NP_033131.2 | 7 - 8 | 821 | 63 |
GenBank mRNA | AK011857.1 | - | 7 - 8 | 714 | 63 |
AK013076.1 | - | 3 - 4 | 292 | 63 | |
AK014749.1 | - | 2 - 3 | 164 | 63 | |
AK135491.1 | - | 8 - 9 | 804 | 63 | |
AK149835.1 | - | 7 - 8 | 691 | 63 | |
AK150011.1 | - | 7 - 8 | 689 | 63 | |
AK152655.1 | - | 7 - 8 | 701 | 63 | |
X63039.1 | - | 7 - 8 | 636 | 63 |