Assay Design



Gene Transcripts

Gene Symbol
Sftpd
Entrez Gene ID
20390
Gene Name
surfactant associated protein D
Gene Aliases
AI573415, SP-D, Sftp4
Chromosome Location
Chr.14: 41172212 - 41185198 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009160.2 NP_033186.1 3 - 4 387 67
GenBank mRNA BC003705.1 - 3 - 4 338 67
L40156.1 - 2 - 3 315 67

View Full Product Details