Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009230.3 | NP_033256.2 | 4 - 5 | 508 | 78 |
GenBank mRNA | AK052299.1 | - | 1 - 2 | 65 | 78 |
AK054149.1 | - | 4 - 5 | 421 | 78 | |
AK142486.1 | - | 4 - 5 | 425 | 78 | |
AK159393.1 | - | 4 - 5 | 438 | 78 | |
AK159529.1 | - | 4 - 5 | 438 | 78 | |
AK160017.1 | - | 4 - 5 | 438 | 78 | |
BC083092.1 | - | 3 - 4 | 322 | 78 | |
DQ903181.1 | - | 4 - 5 | 425 | 78 | |
S81092.1 | - | 4 - 5 | 442 | 78 |