Assay Design



Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
20742
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049
Chromosome Location
Chr.11: 30099395 - 30268204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009260.2 NP_033286.2 13 - 14 3053 66
NM_175836.2 NP_787030.2 13 - 14 2713 66
GenBank mRNA AF017112.1 - 13 - 14 2960 66
AK220456.1 - 14 - 15 2915 66
CF539329.1 - 1 - 2 225 66
M74773.1 - 13 - 14 3033 66

View Full Product Details