Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009260.2 | NP_033286.2 | 25 - 26 | 5965 | 65 |
NM_175836.2 | NP_787030.2 | 25 - 26 | 5625 | 65 | |
GenBank mRNA | AF017112.1 | - | 25 - 26 | 5872 | 65 |
AK161119.1 | - | 4 - 5 | 748 | 65 | |
AK220456.1 | - | 26 - 27 | 5827 | 65 | |
M74773.1 | - | 25 - 26 | 5945 | 65 |