Assay Design



Gene Transcripts

Gene Symbol
Strbp
Entrez Gene ID
20744
Gene Name
spermatid perinuclear RNA binding protein
Gene Aliases
6430510M02Rik, AI852513, C230082I21Rik, C86322, Spnr
Chromosome Location
Chr.2: 37569868 - 37703969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009261.3 NP_033287.2 11 - 12 1506 72
GenBank mRNA BC014710.1 - 12 - 13 1590 72
X84692.1 - - 1443 72

View Full Product Details