Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 5 - 6 | 684 | 73 |
GenBank mRNA | AB006036.1 | - | 5 - 6 | 477 | 73 |
AK136861.1 | - | 5 - 6 | 650 | 73 | |
AK161384.1 | - | 6 - 7 | 743 | 73 | |
AK169580.1 | - | 5 - 6 | 657 | 73 | |
AK178771.1 | - | - | 163 | 73 | |
BC020178.1 | - | 5 - 6 | 581 | 73 | |
BC062941.1 | - | 6 - 7 | 629 | 73 | |
U92456.1 | - | 6 - 7 | 606 | 73 |