Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 14 - 15 | 2071 | 81 |
GenBank mRNA | AB006036.1 | - | 14 - 15 | 1861 | 81 |
AK035962.1 | - | 5 - 6 | 482 | 81 | |
AK041426.1 | - | 4 - 5 | 406 | 81 | |
AK136861.1 | - | 14 - 15 | 2037 | 81 | |
AK149055.1 | - | 3 - 4 | 212 | 81 | |
BC020178.1 | - | 14 - 15 | 1968 | 81 | |
BC062941.1 | - | 14 - 15 | 1990 | 81 |