Assay Design



Gene Transcripts

Gene Symbol
Stra6
Entrez Gene ID
20897
Gene Name
stimulated by retinoic acid gene 6
Gene Aliases
AI891933
Chromosome Location
Chr.9: 58063793 - 58154187 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162475.1 NP_001155947.1 10 - 11 951 70
NM_001162476.1 NP_001155948.1 10 - 11 1262 70
NM_001162479.1 NP_001155951.1 10 - 11 1056 70
NM_009291.2 NP_033317.2 10 - 11 1221 70
GenBank mRNA AF062476.1 - 10 - 11 1221 70
BC075657.1 - 10 - 11 951 70
CF735552.1 - 2 - 3 121 70

View Full Product Details