Assay Design



Gene Transcripts

Gene Symbol
Casq1
Entrez Gene ID
12372
Gene Name
calsequestrin 1
Gene Aliases
CSQ, CSQ-1, CSQ1, sCSQ
Chromosome Location
Chr.1: 172209894 - 172219895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009813.2 NP_033943.2 6 - 7 980 81
GenBank mRNA CR756967.1 - 1 - 2 81
U93291.1 - 6 - 7 936 81

View Full Product Details