Assay Design



Gene Transcripts

Gene Symbol
Casq1
Entrez Gene ID
12372
Gene Name
calsequestrin 1
Gene Aliases
CSQ, CSQ-1, CSQ1, sCSQ
Chromosome Location
Chr.1: 172209894 - 172219895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009813.2 NP_033943.2 7 - 8 1035 69
GenBank mRNA AK049865.1 - 1 - 2 153 69
BC064057.1 - 7 - 8 973 69
U93291.1 - 7 - 8 991 69

View Full Product Details