Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111026.2 | NP_001104496.1 | 8 - 9 | 1625 | 100 |
NM_001111027.2 | NP_001104497.1 | 9 - 10 | 1819 | 100 | |
NM_009822.3 | NP_033952.1 | 8 - 9 | 1557 | 100 | |
GenBank mRNA | AB221624.1 | - | 7 - 8 | 1204 | 100 |
AK032132.1 | - | 8 - 9 | 1492 | 100 | |
AK142006.1 | - | 8 - 9 | 1626 | 100 | |
BC139200.1 | - | 8 - 9 | 1283 | 100 | |
BC139201.1 | - | 8 - 9 | 1287 | 100 | |
D32007.1 | - | 8 - 9 | 1557 | 100 | |
X79989.1 | - | 8 - 9 | 1264 | 100 |