Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281976.1 | NP_001268905.1 | 1 - 2 | 259 | 71 |
NM_009843.4 | NP_033973.2 | 1 - 2 | 259 | 71 | |
GenBank mRNA | AB097214.1 | - | 1 - 2 | 113 | 71 |
AK138891.1 | - | 1 - 2 | 192 | 71 | |
BC042741.1 | - | 1 - 2 | 259 | 71 | |
BC052683.1 | - | 1 - 2 | 179 | 71 | |
X05719.1 | - | 1 - 2 | 170 | 71 |