Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009885.1 | NP_034015.1 | 4 - 5 | 540 | 73 |
GenBank mRNA | AK131818.1 | - | 4 - 5 | 558 | 73 |
AK131824.1 | - | 4 - 5 | 558 | 73 | |
BC006872.1 | - | 4 - 5 | 552 | 73 | |
U33169.1 | - | 4 - 5 | 540 | 73 | |
U37386.1 | - | 4 - 5 | 544 | 73 |